COM448680293 Combi-Blocks, Inc.

4-Morpholin-4-yl-2-phenyl-butyric acid

  • MDL number MFCD09261412
  • Cas Number : 858712-33-1
  • Molecular Weight : 249.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-3337

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-3337
Availability details for
Quantity
  • Availability
  • Add to Cart