COM448680296 Combi-Blocks, Inc.

4-N-Alloc-aminocyclohexanone

  • MDL number MFCD02183577
  • Cas Number : 391248-19-4
  • Molecular Weight : 197.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-3340

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-3340
Availability details for
Quantity
  • Availability
  • Add to Cart