COM448680298 Combi-Blocks, Inc.

6-Chloroimidazo[1,2-a]pyridine-5-carbonitrile

  • MDL number MFCD22887426
  • Cas Number : 1414864-00-8
  • Molecular Weight : 177.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-3347

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-3347
Availability details for
Quantity
  • Availability
  • Add to Cart