COM448680300 Combi-Blocks, Inc.

4-N-Boc-amino-isonipecotic acid methyl ester oxalic acid salt

  • MDL number MFCD17018883
  • Cas Number : 1263094-44-5
  • Molecular Weight : 348.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-3350

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-3350
Availability details for
Quantity
  • Availability
  • Add to Cart