COM448680319 Combi-Blocks, Inc.

4'-N-Piperidinophenyl acetylene

  • MDL number MFCD03839993
  • Cas Number : 41876-66-8
  • Molecular Weight : 185.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-3376

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-3376
Availability details for
Quantity
  • Availability
  • Add to Cart