COM448680322 Combi-Blocks, Inc.

4-Oxo-1-pyridin-4-yl-cyclohexanecarboxylic acid ethyl ester

  • MDL number MFCD17926382
  • Cas Number : 1255098-68-0
  • Molecular Weight : 247.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-3383

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-3383
Availability details for
Quantity
  • Availability
  • Add to Cart