COM448680385 Combi-Blocks, Inc.

5-(2-Amino-ethyl)-2-hydroxy-benzoic acid HBr

  • MDL number MFCD19382506
  • Cas Number : 4900-26-9
  • Molecular Weight : 262.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-3531

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-3531
Availability details for
Quantity
  • Availability
  • Add to Cart