COM448680388 Combi-Blocks, Inc.

5-(2-Methoxyethoxy)-2-nitrobenzoic acid

  • MDL number MFCD09811062
  • Cas Number : 78361-13-4
  • Molecular Weight : 241.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-3534

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-3534
Availability details for
Quantity
  • Availability
  • Add to Cart