COM448680414 Combi-Blocks, Inc.

5-(4-Hydroxy-phenyl)-oxazole-4-carboxylic acid ethyl ester

  • MDL number MFCD02179428
  • Cas Number : 391248-24-1
  • Molecular Weight : 233.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-3585

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-3585
Availability details for
Quantity
  • Availability
  • Add to Cart