COM448680446 Combi-Blocks, Inc.

Cis-2,6-Dimethylmorpholine

  • MDL number MFCD00078428
  • Cas Number : 6485-55-8
  • Molecular Weight : 115.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-3635

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-3635
Availability details for
Quantity
  • Availability
  • Add to Cart