COM448680473 Combi-Blocks, Inc.

5-(5-Chloro-thiophen-2-yl)-[1,3,4]thiadiazol-2-ylamine

  • MDL number MFCD08689595
  • Cas Number : 70057-76-0
  • Molecular Weight : 217.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-3676

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-3676
Availability details for
Quantity
  • Availability
  • Add to Cart