COM448680491 Combi-Blocks, Inc.

5-(Aminomethyl)-2-hydroxy-benzoic acid methyl ester, HCl

  • MDL number MFCD09877748
  • Cas Number : 7383-01-9
  • Molecular Weight : 217.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-3698

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-3698
Availability details for
Quantity
  • Availability
  • Add to Cart