COM448680514 Combi-Blocks, Inc.

5-(Benzyloxy)-2-nitrobenzoic acid

  • MDL number MFCD09728850
  • Cas Number : 61340-15-6
  • Molecular Weight : 273.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-3726

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-3726
Availability details for
Quantity
  • Availability
  • Add to Cart