COM448680616 Combi-Blocks, Inc.

5-(Naphthalen-1-yl)furan-2(3h)-one

  • MDL number MFCD11501457
  • Cas Number : 906560-16-5
  • Molecular Weight : 210.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-3886

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-3886
Availability details for
Quantity
  • Availability
  • Add to Cart