COM448680652 Combi-Blocks, Inc.

5,6,7,8-Tetrahydro-1,8-naphthyridin-2-butylamine

  • MDL number MFCD04115357
  • Cas Number : 380394-88-7
  • Molecular Weight : 205.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-3947

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-3947
Availability details for
Quantity
  • Availability
  • Add to Cart