COM448680703 Combi-Blocks, Inc.

3-(Tetrahydrofuran-2-ylmethoxy)propan-1-amine

  • MDL number MFCD07339549
  • Cas Number : 62035-48-7
  • Molecular Weight : 159.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4073

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4073
Availability details for
Quantity
  • Availability
  • Add to Cart