COM448680713 Combi-Blocks, Inc.

2-Bromo-3-phenylpropanenitrile

  • MDL number MFCD06655784
  • Cas Number : 62448-27-5
  • Molecular Weight : 210.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4094

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4094
Availability details for
Quantity
  • Availability
  • Add to Cart