COM448680864 Combi-Blocks, Inc.

5,7-Dimethoxyquinazolin-4(3h)-one

  • MDL number MFCD13191849
  • Cas Number : 379228-27-0
  • Molecular Weight : 206.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4366

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4366
Availability details for
Quantity
  • Availability
  • Add to Cart