COM448680865 Combi-Blocks, Inc.

1-Boc-5-hydroxypiperidine-3-carboxylic acid

  • MDL number MFCD11656771
  • Cas Number : 1095010-48-2
  • Molecular Weight : 245.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4367

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4367
Availability details for
Quantity
  • Availability
  • Add to Cart