COM448680866 Combi-Blocks, Inc.

5,7-Dimethyl-[1,2,4]triazolo[1,5-a]pyrimidine-2-carbaldehyde

  • MDL number MFCD11846960
  • Cas Number : 55293-96-4
  • Molecular Weight : 176.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4368

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4368
Availability details for
Quantity
  • Availability
  • Add to Cart