COM448680947 Combi-Blocks, Inc.

5-Acetyl-2-tritylamino pyridine

  • MDL number MFCD03839982
  • Cas Number : 49647-11-2
  • Molecular Weight : 378.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4508

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4508
Availability details for
Quantity
  • Availability
  • Add to Cart