COM448681065 Combi-Blocks, Inc.

5-Amino-3-tert-butylisothiazole

  • MDL number MFCD17926249
  • Cas Number : 89151-73-5
  • Molecular Weight : 156.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4682

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4682
Availability details for
Quantity
  • Availability
  • Add to Cart