COM448681071 Combi-Blocks, Inc.

2,3,6-Trichlorobenzoic acid

  • MDL number MFCD00053310
  • Cas Number : 50-31-7
  • Molecular Weight : 225.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4689

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4689
Availability details for
Quantity
  • Availability
  • Add to Cart