COM448681091 Combi-Blocks, Inc.

5-Aminomethyl-dibenzosuberane

  • MDL number MFCD01731070
  • Cas Number : 7351-49-7
  • Molecular Weight : 223.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4728

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4728
Availability details for
Quantity
  • Availability
  • Add to Cart