COM448681108 Combi-Blocks, Inc.

5-Benzyl-octahydro-pyrrolo[3,4-b]pyrrole

  • MDL number MFCD11501198
  • Cas Number : 128740-12-5
  • Molecular Weight : 202.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4752

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4752
Availability details for
Quantity
  • Availability
  • Add to Cart