COM448681109 Combi-Blocks, Inc.

5-Benzyloxy-2,3-dihydro-1h-indole

  • MDL number MFCD07371627
  • Cas Number : 92818-36-5
  • Molecular Weight : 225.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4754

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4754
Availability details for
Quantity
  • Availability
  • Add to Cart