COM448681119 Combi-Blocks, Inc.

5-Bromo-1,4,6-trimethyl-2,3-dihydro-1h-pyrrolo[2,3-b]pyridine

  • MDL number MFCD18711512
  • Cas Number : 627098-12-8
  • Molecular Weight : 241.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4776

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4776
Availability details for
Quantity
  • Availability
  • Add to Cart