COM448681130 Combi-Blocks, Inc.

5-Bromo-2-(2,4-di-tert-butylphenoxy)pyrimidine

  • MDL number MFCD09991552
  • Cas Number : 1017788-99-6
  • Molecular Weight : 363.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4802

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4802
Availability details for
Quantity
  • Availability
  • Add to Cart