COM448681137 Combi-Blocks, Inc.

5-Bromo-2-(2-methoxy-4-methylphenoxy)pyrimidine

  • MDL number MFCD09991554
  • Cas Number : 1017789-03-5
  • Molecular Weight : 295.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4814

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4814
Availability details for
Quantity
  • Availability
  • Add to Cart