COM448681141 Combi-Blocks, Inc.

5-Bromo-2-(methylsulfanyl)pyridine-3-carboxylic acid

  • MDL number MFCD16159775
  • Cas Number : 1220422-10-5
  • Molecular Weight : 248.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4821

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4821
Availability details for
Quantity
  • Availability
  • Add to Cart