COM448681142 Combi-Blocks, Inc.

5-Bromo-2-(m-tolyloxy)pyrimidine

  • MDL number MFCD03646352
  • Cas Number : 73221-74-6
  • Molecular Weight : 265.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4822

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4822
Availability details for
Quantity
  • Availability
  • Add to Cart