COM448681144 Combi-Blocks, Inc.

5-Bromo-2-(o-tolyloxy)pyrimidine

  • MDL number MFCD09991551
  • Cas Number : 73267-74-0
  • Molecular Weight : 265.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4825

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4825
Availability details for
Quantity
  • Availability
  • Add to Cart