COM448681149 Combi-Blocks, Inc.

5-Bromo-2,3-dihydro-3,3-dimethylbenzofuran

  • MDL number MFCD18374741
  • Cas Number : 68505-84-0
  • Molecular Weight : 227.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4832

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4832
Availability details for
Quantity
  • Availability
  • Add to Cart