COM448681156 Combi-Blocks, Inc.

5-Bromo-2-isopropylisoindolin-1-one

  • MDL number MFCD12400939
  • Cas Number : 864866-47-7
  • Molecular Weight : 254.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4841

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4841
Availability details for
Quantity
  • Availability
  • Add to Cart