COM448681173 Combi-Blocks, Inc.

5-Bromo-6,7-dihydro-benzo[b]thiophen-4(5h)-one

  • MDL number MFCD08694564
  • Cas Number : 2513-49-7
  • Molecular Weight : 231.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4872

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4872
Availability details for
Quantity
  • Availability
  • Add to Cart