COM448681184 Combi-Blocks, Inc.

5-Bromo-n-(2,2-dimethoxyethyl)pyrimidin-2-amine

  • MDL number MFCD03646021
  • Cas Number : 885267-37-8
  • Molecular Weight : 262.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4892

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4892
Availability details for
Quantity
  • Availability
  • Add to Cart