COM448681203 Combi-Blocks, Inc.

5-Chloro-3-methylthieno[3,2-b]pyridine

  • MDL number MFCD21607352
  • Cas Number : 1356016-33-5
  • Molecular Weight : 183.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4945

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4945
Availability details for
Quantity
  • Availability
  • Add to Cart