COM448681204 Combi-Blocks, Inc.

5-Chloro-4-fluoro-1h-indole-2-carboxylic acid

  • MDL number MFCD09835295
  • Cas Number : 186446-26-4
  • Molecular Weight : 213.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4946

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4946
Availability details for
Quantity
  • Availability
  • Add to Cart