COM448681206 Combi-Blocks, Inc.

5-Chloroacetyl-2,3-dihydrobenzofuran

  • MDL number MFCD03618417
  • Cas Number : 64089-34-5
  • Molecular Weight : 196.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4948

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4948
Availability details for
Quantity
  • Availability
  • Add to Cart