COM448681211 Combi-Blocks, Inc.

5-Chloroimidazol[1,2-a]pyrimidine

  • MDL number MFCD10698094
  • Cas Number : 944896-82-6
  • Molecular Weight : 153.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4956

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4956
Availability details for
Quantity
  • Availability
  • Add to Cart