COM448681212 Combi-Blocks, Inc.

5-Chloromethyl-3-(4-chloro-3-nitro-phenyl)-[1,2,4]oxadiazole

  • MDL number MFCD07369936
  • Cas Number : 187399-90-2
  • Molecular Weight : 274.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4958

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4958
Availability details for
Quantity
  • Availability
  • Add to Cart