COM448681213 Combi-Blocks, Inc.

5-Chloromethyl-3-(4-methylsulfanyl-phenyl)-[1,2,4]oxadiazole

  • MDL number MFCD07369935
  • Cas Number : 845887-08-3
  • Molecular Weight : 240.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4959

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4959
Availability details for
Quantity
  • Availability
  • Add to Cart