COM448681223 Combi-Blocks, Inc.

5-Cyanoindole-1-acetic acid

  • MDL number MFCD03839835
  • Cas Number : 202124-67-2
  • Molecular Weight : 200.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4980

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4980
Availability details for
Quantity
  • Availability
  • Add to Cart