COM448681317 Combi-Blocks, Inc.

5-Oxo-[1,4]diazepane-1-carboxylic acid benzyl ester

  • MDL number MFCD03426387
  • Cas Number : 18158-16-2
  • Molecular Weight : 248.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-5196

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-5196
Availability details for
Quantity
  • Availability
  • Add to Cart