COM448681347 Combi-Blocks, Inc.

6-(4-Methoxyphenyl)morpholin-3-one

  • MDL number MFCD09832087
  • Cas Number : 5196-94-1
  • Molecular Weight : 207.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-5260

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-5260
Availability details for
Quantity
  • Availability
  • Add to Cart