COM448681367 Combi-Blocks, Inc.

6,7-Difluoro-4-oxo-1,4-dihydroquinoline-3-carbonitrile

  • MDL number MFCD00498955
  • Cas Number : 957137-97-2
  • Molecular Weight : 206.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-5298

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-5298
Availability details for
Quantity
  • Availability
  • Add to Cart