COM448681368 Combi-Blocks, Inc.

6,7-Dihydro-5h-[1,3]dioxolo[4,5-f]isoindole, HCl

  • MDL number MFCD17926273
  • Cas Number : 1255099-33-2
  • Molecular Weight : 199.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-5299

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-5299
Availability details for
Quantity
  • Availability
  • Add to Cart