COM448681391 Combi-Blocks, Inc.

6,7-Dimethyl-1h-benzo[d]imidazole-2-thiol

  • MDL number MFCD09788614
  • Cas Number : 116643-17-5
  • Molecular Weight : 178.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-5344

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-5344
Availability details for
Quantity
  • Availability
  • Add to Cart