COM448681392 Combi-Blocks, Inc.

6,9-Diaza-spiro[4.5]decane DiHCl

  • MDL number MFCD06796600
  • Cas Number : 145122-55-0
  • Molecular Weight : 213.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-5348

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-5348
Availability details for
Quantity
  • Availability
  • Add to Cart