COM448681393 Combi-Blocks, Inc.

6-Acetyaminochroman-4-one

  • MDL number MFCD06202343
  • Cas Number : 103646-29-3
  • Molecular Weight : 205.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-5354

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-5354
Availability details for
Quantity
  • Availability
  • Add to Cart